Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2410127L17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026120.4 | NP_080396.2 | 1, 2 | 434 | ||
GenBank mRNA | AI509964.1 | - | 1, 2 | 220 | ||
AK010784.1 | - | 1, 2 | 319 | |||
AK178550.1 | - | 1, 2 | 118 | |||
AK182869.1 | - | 1, 2 | 118 | |||
BB656580.1 | - | 1, 2 | 450 | |||
BC019559.1 | - | 1, 2 | 624 | |||
BC043335.1 | - | 1, 2 | 388 | 5030742 | ||
BC092281.1 | - | 1, 2 | 266 | 4950426 |