Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD51AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006479.4 | NP_006470.1 | 3, 4 | 373 | ||
GenBank mRNA | AF006259.1 | - | 3, 4 | 268 | ||
AK291948.1 | - | 3, 4 | 267 | |||
BC006992.1 | - | 3, 4 | 267 | 3997352 | ||
DC397655.1 | - | 3, 4 | 373 |