Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhdds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026144.4 | NP_080420.2 | 6 | 631 | ||
GenBank mRNA | AK014317.1 | - | 7 | 819 | ||
AK017556.1 | - | 6 | 703 | |||
AK034277.1 | - | 6 | 693 | |||
AK036929.1 | - | 6 | 709 | |||
AK049991.1 | - | 6 | 632 | |||
AK135087.1 | - | 6 | 693 | |||
AK149138.1 | - | 6 | 663 | |||
AK171207.1 | - | 6 | 705 | |||
BC004011.1 | - | 6 | 679 | 3493585 |