Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001996.3 | NP_001987.2 | 2 | 299 | ||
NM_006485.3 | NP_006476.2 | 2 | 299 | |||
NM_006486.2 | NP_006477.2 | 2 | 299 | |||
NM_006487.2 | NP_006478.2 | 2 | 299 | |||
GenBank mRNA | AF126110.1 | - | 2 | 162 | ||
AF217999.1 | - | 2 | 241 | |||
AK075460.1 | - | 2 | 255 | |||
AK075566.1 | - | 2 | 258 | |||
AK300806.1 | - | 2 | 235 | |||
AK316291.1 | - | 2 | 235 | |||
BC022497.1 | - | 2 | 255 | 4819750 | ||
BX424750.2 | - | 2 | 297 | |||
U01244.1 | - | 2 | 162 | |||
X53741.1 | - | 2 | 162 | |||
X53742.1 | - | 2 | 162 | |||
X53743.1 | - | 2 | 162 |