Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286560.1 | NP_001273489.1 | 7, 8 | 914 | ||
NM_026168.4 | NP_080444.1 | 8, 9 | 1023 | |||
NM_026355.3 | NP_080631.1 | 8, 9 | 1023 | |||
GenBank mRNA | AK004661.1 | - | 8, 9 | 687 | ||
AK007498.1 | - | 8, 9 | 672 | |||
AK008971.1 | - | 8, 9 | 679 | |||
AK009483.1 | - | 8, 9 | 675 | |||
AK010529.1 | - | 8, 9 | 672 | |||
AK016275.1 | - | 8, 9 | 1024 | |||
AK017876.1 | - | 8, 9 | 707 | |||
AK030045.1 | - | 8, 9 | 673 | |||
AK086727.1 | - | 8, 9 | 675 | |||
AK135968.1 | - | 5, 6 | 360 | |||
AK162267.1 | - | 7, 8 | 647 | |||
AK167391.1 | - | 8, 9 | 676 | |||
BC006895.1 | - | 8, 9 | 641 | 2651322 | ||
BC018188.1 | - | 8, 9 | 675 | 3711066 | ||
BC026558.1 | - | 8, 9 | 635 | 5030661 | ||
BC064749.1 | - | 8, 9 | 736 | 30046220 |