Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286560.1 | NP_001273489.1 | 9 | 1013 | ||
NM_026168.4 | NP_080444.1 | 10 | 1122 | |||
NM_026355.3 | NP_080631.1 | 10 | 1122 | |||
GenBank mRNA | AK004661.1 | - | 10 | 786 | ||
AK007498.1 | - | 10 | 771 | |||
AK008971.1 | - | 10 | 778 | |||
AK009483.1 | - | 10 | 774 | |||
AK010529.1 | - | 10 | 771 | |||
AK016275.1 | - | 10 | 1123 | |||
AK017876.1 | - | 10 | 806 | |||
AK030045.1 | - | 10 | 772 | |||
AK086727.1 | - | 10 | 774 | |||
AK135968.1 | - | 7 | 459 | |||
AK162267.1 | - | 9 | 746 | |||
AK167391.1 | - | 10 | 775 | |||
BC006895.1 | - | 10 | 740 | 2651322 | ||
BC018188.1 | - | 10 | 774 | 3711066 | ||
BC026558.1 | - | 10 | 734 | 5030661 | ||
BC064749.1 | - | 10 | 835 | 30046220 |