Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026174.3 | NP_080450.1 | 3 | 333 | ||
GenBank mRNA | AB093238.1 | - | 3 | 275 | ||
AK004761.1 | - | 3 | 300 | |||
AK042053.1 | - | 2 | 1774 | |||
AK192189.1 | - | 2 | 206 | |||
AK193291.1 | - | 2 | 205 | |||
AK197306.1 | - | 2 | 205 | |||
AK197494.1 | - | 2 | 206 | |||
AK198046.1 | - | 2 | 205 | |||
AK201574.1 | - | 2 | 209 | |||
AK204612.1 | - | 2 | 205 | |||
AK207715.1 | - | 2 | 226 | |||
AK213995.1 | - | 2 | 205 | |||
BC006924.1 | - | 1 | 82 | |||
BC043134.1 | - | 3 | 285 | 6811715 | ||
BC052090.1 | - | 3 | 286 | |||
BY276189.1 | - | 3 | 335 |