Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026174.3 | NP_080450.1 | 4 | 537 | ||
GenBank mRNA | AB093238.1 | - | 4 | 479 | ||
AK004761.1 | - | 4 | 504 | |||
AK045342.1 | - | 516 | ||||
BC006924.1 | - | 2 | 286 | |||
BC043134.1 | - | 4 | 489 | 6811715 |