Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026178.2 | NP_080454.1 | 7 | 888 | ||
GenBank mRNA | AK004819.1 | - | 7 | 842 | ||
AK011418.1 | - | 7 | 853 | |||
AK158577.1 | - | 7 | 846 | |||
AK171637.1 | - | 7 | 842 | |||
AY424300.1 | - | 7 | 728 | |||
BC021914.1 | - | 7 | 888 | 4506710 | ||
BC034117.1 | - | 8 | 1240 |