Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286005.1 | NP_001272934.1 | 3, 4 | 429 | ||
NM_026180.3 | NP_080456.1 | 4, 5 | 681 | |||
GenBank mRNA | AF324495.1 | - | 4, 5 | 671 | ||
AK004871.1 | - | 4, 5 | 638 | |||
AK050938.1 | - | 3, 4 | 431 | |||
AY196215.1 | - | 4, 5 | 668 | |||
AY196216.1 | - | 4, 5 | 668 | |||
BC138484.1 | - | 4, 5 | 645 | 8861504 | ||
BC145293.1 | - | 4, 5 | 645 | 9053847 |