Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026194.4 | NP_080470.2 | 2 | 209 | ||
GenBank mRNA | AK007993.2 | - | 2 | 154 | ||
AK032663.1 | - | 2 | 145 | |||
AK076206.1 | - | 2 | 154 | |||
AK129211.1 | - | 2 | 151 | |||
AK132697.1 | - | 2 | 173 | |||
BC058809.1 | - | 2 | 148 | |||
BC080203.1 | - | 2 | 187 | |||
BC099529.1 | - | 2 | 197 | |||
BC132195.1 | - | 2 | 161 | 40130472 | ||
BC138153.1 | - | 2 | 161 | 8861172 |