Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303245.1 | NP_001290174.1 | 2 | 255 | ||
NM_001303246.1 | NP_001290175.1 | 2 | 255 | |||
NM_006506.3 | NP_006497.2 | 2 | 255 | |||
XM_011513059.2 | XP_011511361.1 | 3 | 1404 | |||
XM_017006968.1 | XP_016862457.1 | 3 | 388 | |||
XM_017006969.1 | XP_016862458.1 | 3 | 387 | |||
XM_017006970.1 | XP_016862459.1 | 3 | 394 | |||
XM_017006971.1 | XP_016862460.1 | 3 | 396 | |||
XM_017006972.1 | XP_016862461.1 | 1 | 85 | |||
GenBank mRNA | AF115573.1 | - | 2 | 218 | ||
AK292016.1 | - | 2 | 253 | |||
D78155.1 | - | 2 | 218 | |||
D82880.1 | - | 2 | 218 | |||
DA792257.1 | - | 2 | 221 |