Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026213.3 | NP_080489.1 | 5 | 841 | ||
GenBank mRNA | AK010758.1 | - | 5 | 659 | ||
AK013464.1 | - | 5 | 687 | |||
AK050426.1 | - | 5 | 681 | |||
AK075643.1 | - | 5 | 667 | |||
BC017533.1 | - | 4 | 606 | |||
BC061018.1 | - | 5 | 841 | 6590550 |