Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026214.3 | NP_080490.3 | 2 | 1008 | ||
GenBank mRNA | AK008741.1 | - | 2 | 1003 | ||
AK017592.1 | - | 1 | 739 | |||
AK032314.1 | - | 1 | 715 | |||
AK032624.1 | - | 1 | 746 | |||
BC028748.1 | - | 1 | 723 | 1332429 |