Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYSLTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282186.1 | NP_001269115.1 | 2 | 681 | ||
NM_001282187.1 | NP_001269116.1 | 4 | 851 | |||
NM_001282188.1 | NP_001269117.1 | 4 | 828 | |||
NM_006639.3 | NP_006630.1 | 3 | 768 | |||
GenBank mRNA | AF119711.1 | - | 1 | 267 | ||
AF133266.1 | - | 1 | 267 | |||
AK313643.1 | - | 3 | 692 | |||
BC035750.1 | - | 3 | 560 | 5750221 |
Set Membership: |
G-protein coupled receptor |