Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026231.2 | NP_080507.2 | 6 | 1468 | ||
GenBank mRNA | AK013967.1 | - | 3 | 320 | ||
AK017059.1 | - | 4 | 743 | |||
AK020021.1 | - | 5 | 1078 | |||
AK029724.1 | - | 6 | 1467 | |||
AK033409.1 | - | 6 | 1506 | |||
AK044441.1 | - | 6 | 1476 | |||
BC047136.1 | - | 6 | 1468 | 6311866 | ||
BC061240.1 | - | 6 | 1464 | 6743615 |