Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026231.2 | NP_080507.2 | 7, 8 | 1594 | ||
GenBank mRNA | AK013967.1 | - | 4, 5 | 446 | ||
AK017059.1 | - | 5, 6 | 869 | |||
AK020021.1 | - | 6, 7 | 1204 | |||
AK029724.1 | - | 7, 8 | 1594 | |||
AK033409.1 | - | 7, 8 | 1632 | |||
AK044441.1 | - | 7, 8 | 1602 | |||
BC047136.1 | - | 7, 8 | 1594 | 6311866 | ||
BC061240.1 | - | 7, 8 | 1590 | 6743615 |