Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026234.4 | NP_080510.1 | 1 | 319 | ||
GenBank mRNA | AK018560.1 | - | 1 | 272 | ||
AK040397.1 | - | 1 | 295 | |||
AK044641.1 | - | 1 | 289 | |||
AK080242.1 | - | 1 | 280 | |||
AK082746.1 | - | 1 | 317 | |||
AK088117.1 | - | 1 | 299 | |||
AK148197.1 | - | 1 | 255 | |||
BC005650.1 | - | 1 | 304 | 3257474 | ||
BC083115.1 | - | 1 | 295 | 5364341 |