Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Patl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026251.2 | NP_080527.2 | 9 | 1056 | ||
GenBank mRNA | AK010811.1 | - | 2 | 128 | ||
AK015195.1 | - | 9 | 1057 | |||
BC145646.1 | - | 7 | 837 | 40130949 | ||
DV658440.1 | - | 7 | 667 |