Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006520.2 | NP_006511.1 | 5 | 1097 | ||
GenBank mRNA | BC000968.2 | - | 5 | 1025 | 3448623 | |
U02556.1 | - | 5 | 1036 |