Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026267.2 | NP_080543.2 | 8 | 1269 | ||
GenBank mRNA | AK004805.1 | - | 8 | 1270 | ||
AK005263.1 | - | 8 | 1260 | |||
AK081598.1 | - | 8 | 1270 | |||
AK159237.1 | - | 1 | 379 | |||
AK168500.1 | - | 8 | 1270 | |||
AK220236.1 | - | 8 | 1253 |