Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026274.4 | NP_080550.3 | 2 | 918 | ||
GenBank mRNA | AK032416.1 | - | 2 | 881 | ||
AK076799.1 | - | 2 | 491 | |||
AK141443.1 | - | 2 | 468 | |||
AK146028.1 | - | 2 | 882 | |||
AK151018.1 | - | 2 | 888 | |||
AK152024.1 | - | 2 | 888 | |||
AK166167.1 | - | 2 | 893 | |||
AK172567.1 | - | 2 | 884 | |||
AK173313.1 | - | 2 | 537 | |||
BC010833.1 | - | 2 | 533 | 4235165 | ||
BC054121.1 | - | 2 | 904 | 6393910 |