Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026274.4 | NP_080550.3 | 4 | 1122 | ||
GenBank mRNA | AK032416.1 | - | 4 | 1085 | ||
AK076799.1 | - | 4 | 695 | |||
AK141443.1 | - | 4 | 672 | |||
AK150941.1 | - | 3 | 597 | |||
AK151018.1 | - | 4 | 1092 | |||
AK152024.1 | - | 4 | 1092 | |||
AK152435.1 | - | 3 | 597 | |||
AK172567.1 | - | 4 | 1088 | |||
AK173313.1 | - | 4 | 741 | |||
BC010833.1 | - | 4 | 737 | 4235165 | ||
BC054121.1 | - | 4 | 1108 | 6393910 |