Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026275.4 | NP_080551.1 | 5 | 662 | ||
GenBank mRNA | AJ240086.1 | - | 5 | 621 | ||
AK003550.1 | - | 5 | 682 | |||
AK007517.1 | - | 5 | 796 | |||
AK075703.1 | - | 5 | 672 | |||
AK075714.1 | - | 5 | 663 | |||
BC011112.1 | - | 5 | 670 | 3982808 |