Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bend5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026279.3 | NP_080555.1 | 2, 3 | 522 | ||
GenBank mRNA | AK014057.1 | - | 2, 3 | 512 | ||
AK075885.1 | - | 2, 3 | 522 | |||
AK167425.1 | - | 2, 3 | 342 | |||
BC037633.1 | - | 2, 3 | 355 |