Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026302.3 | NP_080578.1 | 1 | 123 | ||
GenBank mRNA | AK003238.1 | - | 1 | 116 | ||
AK016059.1 | - | 1 | 177 | |||
AK019382.1 | - | 1 | 125 | |||
AK034292.1 | - | 1 | 160 | |||
AK050896.1 | - | 1 | 127 | |||
AK089264.1 | - | 1 | 117 | |||
AK141855.1 | - | 1 | 124 | |||
AK160813.1 | - | 1 | 129 | |||
AK162552.1 | - | 1 | 130 | |||
AK163240.1 | - | 1 | 145 | |||
AK168484.1 | - | 1 | 129 | |||
AK197489.1 | - | 1 | 23 | |||
BC006677.1 | - | 1 | 86 | |||
BC034725.1 | - | 1 | 123 | 3667031 |