Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: l7Rn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291286.1 | NP_001278215.1 | 3 | 509 | ||
NM_001291287.1 | NP_001278216.1 | 4 | 573 | |||
NM_001291288.1 | NP_001278217.1 | 4 | 650 | |||
NM_001291289.1 | NP_001278218.1 | 2 | 271 | |||
NM_026304.3 | NP_080580.1 | 3 | 509 | |||
GenBank mRNA | AK002306.1 | - | 3 | 491 | ||
AK003324.1 | - | 2 | 262 | |||
AK008604.1 | - | 2 | 273 | |||
AK012166.1 | - | 3 | 475 | |||
AK206361.1 | - | 2 | 284 | |||
BC003916.1 | - | 3 | 457 | 3485003 | ||
BC083330.1 | - | 4 | 511 | |||
DQ196315.1 | - | 3 | 487 | |||
DQ196316.1 | - | 3 | 487 |