Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166370.1 | NP_001159842.1 | 2, 3 | 674 | ||
NM_026306.3 | NP_080582.3 | 3, 4 | 576 | |||
GenBank mRNA | AK002791.1 | - | 2, 3 | 221 | ||
AK003292.1 | - | 2, 3 | 219 | |||
BC016191.1 | - | 2, 3 | 596 | 4483787 | ||
BC019418.1 | - | 2, 3 | 208 | 4162006 | ||
BC087959.1 | - | 3, 4 | 576 | 6766768 | ||
DV654053.1 | - | 2, 3 | 564 |