Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166370.1 | NP_001159842.1 | 4 | 874 | ||
NM_026306.3 | NP_080582.3 | 5 | 776 | |||
GenBank mRNA | AK002791.1 | - | 4 | 421 | ||
AK003292.1 | - | 4 | 419 | |||
BC016191.1 | - | 4 | 793 | 4483787 | ||
BC019418.1 | - | 3 | 483 | 4162006 | ||
BC087959.1 | - | 5 | 776 | 6766768 |