Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026307.3 | NP_080583.3 | 7 | 598 | ||
NM_026948.3 | NP_081224.1 | 6 | 546 | |||
GenBank mRNA | AK002828.1 | - | 7 | 542 | ||
AK003381.1 | - | 6 | 486 | |||
AK003647.1 | - | 7 | 540 | |||
AK004197.1 | - | 7 | 540 | |||
AK007580.1 | - | 5 | 675 | |||
AK012608.1 | - | 4 | 1018 | |||
AK159336.1 | - | 7 | 545 | |||
AK199328.1 | - | 4 | 205 | |||
AK207031.1 | - | 4 | 205 | |||
AK207668.1 | - | 3 | 280 | |||
BC021463.1 | - | 5 | 660 | |||
BC024422.1 | - | 7 | 528 | 5005082 | ||
BC028531.1 | - | 6 | 637 | |||
BC037500.1 | - | 4 | 628 | |||
CA323205.1 | - | 6 | 511 | |||
CK624897.1 | - | 6 | 523 |