Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyx1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026314.3 | NP_080590.3 | 4 | 560 | ||
GenBank mRNA | AY428507.1 | - | 4 | 560 | ||
BC026462.1 | - | 4 | 517 | 4483015 |