Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007225.1 | NP_001007226.1 | 2, 3 | 326 | ||
NM_001291869.1 | NP_001278798.1 | 2, 3 | 326 | |||
NM_006548.4 | NP_006539.3 | 2, 3 | 326 | |||
XM_017005558.1 | XP_016861047.1 | 2, 3 | 331 | |||
XM_017005559.1 | XP_016861048.1 | 2, 3 | 332 | |||
XM_017005560.1 | XP_016861049.1 | 2, 3 | 228 | |||
XM_017005561.1 | XP_016861050.1 | 2, 3 | 333 | |||
XM_017005562.1 | XP_016861051.1 | 2, 3 | 332 | |||
XM_017005563.1 | XP_016861052.1 | 2, 3 | 332 | |||
XM_017005564.1 | XP_016861053.1 | 2, 3 | 332 | |||
GenBank mRNA | AF117107.1 | - | 2, 3 | 293 | ||
AK290664.1 | - | 2, 3 | 327 | |||
AK309611.1 | - | 2, 3 | 280 | |||
BC021290.2 | - | 2, 3 | 311 | 5090334 |