Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hypk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026318.3 | NP_080594.2 | 1, 2 | 211 | ||
GenBank mRNA | AK003580.1 | - | 1, 2 | 205 | ||
AK009109.1 | - | 1, 2 | 199 | |||
AK168506.1 | - | 1, 2 | 180 | |||
BC021588.1 | - | 1, 2 | 175 | 5116361 | ||
BC038469.1 | - | 1, 2 | 213 | 5028838 | ||
BY165803.1 | - | 1, 2 | 213 |