Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hypk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026318.3 | NP_080594.2 | 4 | 392 | ||
GenBank mRNA | AK003580.1 | - | 4 | 386 | ||
AK009109.1 | - | 4 | 380 | |||
AK168506.1 | - | 4 | 361 | |||
BC021588.1 | - | 3 | 646 | 5116361 | ||
BC038469.1 | - | 4 | 394 | 5028838 | ||
BC060244.1 | - | 3 | 1141 |