Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kirrel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190911.1 | NP_001177840.1 | 17 | 3624 | ||
NM_001190912.1 | NP_001177841.1 | 16 | 3621 | |||
NM_001190914.1 | NP_001177843.1 | 17 | 3689 | |||
NM_026324.3 | NP_080600.1 | 16 | 3588 | |||
GenBank mRNA | AK005197.1 | - | 1 | 1034 | ||
AK045373.1 | - | 17 | 3529 | |||
AK079332.1 | - | 17 | 3628 | |||
AK171866.1 | - | 13 | 2939 | |||
AY169782.1 | - | 16 | 3588 | |||
BC063072.1 | - | 16 | 3416 | 5695003 |