Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam114a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168667.1 | NP_001162138.1 | 2 | 224 | ||
NM_001168668.1 | NP_001162139.1 | 2 | 224 | |||
NM_026342.3 | NP_080618.1 | 2 | 227 | |||
GenBank mRNA | AK007968.1 | - | 2 | 201 | ||
AK018570.1 | - | 2 | 200 | |||
AK076316.1 | - | 2 | 198 | |||
AK149763.1 | - | 2 | 202 | |||
BC019518.1 | - | 2 | 184 | 4456429 | ||
BY134675.1 | - | 2 | 224 |