Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mansc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026345.4 | NP_080621.1 | 3 | 651 | ||
GenBank mRNA | AK002644.1 | - | 3 | 560 | ||
AK018635.1 | - | 3 | 542 | |||
AK018660.1 | - | 3 | 564 | |||
AK033526.1 | - | 3 | 563 | |||
AK033557.1 | - | 3 | 652 | |||
AK046837.1 | - | 3 | 540 | |||
AK171842.1 | - | 3 | 568 | |||
BC039930.1 | - | 3 | 522 | 3598325 |