Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YKT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006555.3 | NP_006546.1 | 2 | 276 | ||
XM_005249582.4 | XP_005249639.1 | 2 | 277 | |||
XM_017011682.1 | XP_016867171.1 | 2 | 287 | |||
GenBank mRNA | AB451317.1 | - | 2 | 119 | ||
AB451447.1 | - | 2 | 119 | |||
AF091079.1 | - | 2 | 179 | |||
AK223488.1 | - | 2 | 233 | |||
AK299158.1 | - | 2 | 287 | |||
AK312622.1 | - | 2 | 265 | |||
AU130339.1 | - | 2 | 287 | |||
BC007319.2 | - | 2 | 206 | 2989381 | ||
BT007078.1 | - | 2 | 119 | |||
CR456989.1 | - | 2 | 119 | |||
CR542008.1 | - | 2 | 119 | |||
U95735.1 | - | 2 | 178 |