Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loh12cr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170479.1 | NP_001163950.1 | 4 | 1339 | ||
NM_026371.3 | NP_080647.2 | 4 | 1481 | |||
GenBank mRNA | AK007423.1 | - | 4 | 1338 | ||
AK018019.1 | - | 4 | 1397 | |||
AK151342.1 | - | 4 | 1347 | |||
AK169615.1 | - | 4 | 1353 | |||
BC023059.1 | - | 4 | 1440 | 5355560 |