Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026380.3 | NP_080656.2 | 7 | 1772 | ||
GenBank mRNA | AK044337.1 | - | 7 | 1646 | ||
AK142301.1 | - | 7 | 1641 | |||
BC065056.1 | - | 7 | 1705 | 5705033 | ||
BC094577.1 | - | 6 | 1388 | 6852696 |