Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026403.3 | NP_080679.3 | 3 | 804 | ||
GenBank mRNA | AK011570.1 | - | 3 | 794 | ||
AK015585.1 | - | 3 | 554 | |||
AK032873.1 | - | 3 | 788 | |||
AK037831.1 | - | 3 | 792 | |||
AK050912.1 | - | 3 | 795 | |||
AK083861.1 | - | 3 | 673 | |||
AK139689.1 | - | 3 | 755 | |||
AK146020.1 | - | 3 | 759 | |||
AK146687.1 | - | 3 | 776 | |||
AK152732.1 | - | 3 | 778 | |||
AK154387.1 | - | 3 | 796 | |||
AK159798.1 | - | 3 | 778 | |||
BC018523.1 | - | 3 | 784 | 3499986 |