Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026403.3 | NP_080679.3 | 10 | 2130 | ||
GenBank mRNA | AK011570.1 | - | 10 | 2120 | ||
AK032873.1 | - | 10 | 2114 | |||
AK050912.1 | - | 10 | 2121 | |||
AK083861.1 | - | 10 | 1999 | |||
AK139689.1 | - | 10 | 2083 | |||
AK146020.1 | - | 10 | 2085 | |||
AK146687.1 | - | 10 | 2102 | |||
AK152732.1 | - | 10 | 2104 | |||
AK154387.1 | - | 10 | 2122 | |||
AK159798.1 | - | 10 | 2104 | |||
AK208092.1 | - | 1 | 168 | |||
BC018523.1 | - | 10 | 2110 | 3499986 |