Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026405.3 | NP_080681.1 | 2, 3 | 639 | ||
GenBank mRNA | AB232627.1 | - | 2, 3 | 520 | ||
AK012701.1 | - | 2, 3 | 651 | |||
AK078874.1 | - | 2, 3 | 331 | |||
AK088661.1 | - | 2, 3 | 640 | |||
AK151543.1 | - | 2, 3 | 613 | |||
AK159102.1 | - | 2, 3 | 666 | |||
AK159901.1 | - | 2, 3 | 666 | |||
AK163620.1 | - | 2, 3 | 590 | |||
AK180112.1 | - | 1, 2 | 154 | |||
AK206948.1 | - | 2, 3 | 349 | |||
AK216759.1 | - | 1, 2 | 279 | |||
AY135650.1 | - | 2, 3 | 544 | |||
BC016409.1 | - | 557 | 4456902 | |||
BC055945.1 | - | 565 | 4161169 |