Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncaip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199151.1 | NP_001186080.1 | 4 | 482 | ||
NM_001199153.1 | NP_001186082.1 | 4 | 387 | |||
NM_001199154.1 | NP_001186083.1 | 4 | 657 | |||
NM_026408.4 | NP_080684.3 | 4 | 482 | |||
GenBank mRNA | AF348447.1 | - | 4 | 410 | ||
AK013040.1 | - | 2 | 215 | |||
AK016941.1 | - | 4 | 399 | |||
AK044974.1 | - | 4 | 680 | |||
AK081942.1 | - | 4 | 659 | |||
AK082079.1 | - | 4 | 486 | |||
AK132346.1 | - | 4 | 585 | |||
AK160111.1 | - | 4 | 486 | |||
BC098489.1 | - | 4 | 386 | 30096466 | ||
BC130225.1 | - | 4 | 372 | |||
BC152544.1 | - | 4 | 370 |