Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190795.1 | NP_001177724.1 | 2 | 192 | ||
NM_026409.4 | NP_080685.2 | 2 | 192 | |||
GenBank mRNA | AK012782.1 | - | 2 | 172 | ||
AK017554.1 | - | 2 | 134 | |||
AK033741.1 | - | 2 | 169 | |||
AK077525.1 | - | 2 | 173 | |||
AK129402.1 | - | 2 | 170 | |||
AK154422.1 | - | 2 | 193 | |||
AK167149.1 | - | 2 | 187 | |||
BC043052.1 | - | 2 | 142 | |||
BC117786.1 | - | 2 | 154 | 40092124 | ||
BC117787.1 | - | 2 | 154 | 40092125 | ||
BY202366.1 | - | 2 | 193 |