Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190795.1 | NP_001177724.1 | 9 | 994 | ||
NM_026409.4 | NP_080685.2 | 9 | 994 | |||
GenBank mRNA | AK012782.1 | - | 9 | 974 | ||
AK017554.1 | - | 9 | 936 | |||
AK033741.1 | - | 9 | 971 | |||
AK077525.1 | - | 9 | 975 | |||
AK129402.1 | - | 9 | 972 | |||
AK154422.1 | - | 9 | 995 | |||
AK167149.1 | - | 8 | 2989 | |||
BC043052.1 | - | 9 | 944 | |||
BC117786.1 | - | 9 | 956 | 40092124 | ||
BC117787.1 | - | 9 | 956 | 40092125 |