Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026417.4 | NP_080693.2 | 6 | 1088 | ||
GenBank mRNA | AK009618.1 | - | 6 | 1079 | ||
AK017721.1 | - | 6 | 1064 | |||
AK017722.1 | - | 6 | 1067 | |||
AK083301.1 | - | 6 | 1068 | |||
AK159227.1 | - | 6 | 1048 | |||
AK166347.1 | - | 6 | 1076 | |||
BC003936.1 | - | 7 | 1263 | 3486503 |