Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enoph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163035.1 | NP_001156507.1 | 4, 5 | 746 | ||
NM_026421.3 | NP_080697.2 | 4, 5 | 746 | |||
GenBank mRNA | AK009954.1 | - | 4, 5 | 729 | ||
AK079062.1 | - | 4, 5 | 740 | |||
AK080714.1 | - | 4, 5 | 697 | |||
AK082747.1 | - | 4, 5 | 721 | |||
AK152021.1 | - | 4, 5 | 721 | |||
AK159356.1 | - | 4, 5 | 719 | |||
AK167575.1 | - | 4, 5 | 757 | |||
AK168696.1 | - | 4, 5 | 615 | |||
AK177339.1 | - | 3, 4 | 357 | |||
BC021429.1 | - | 4, 5 | 673 | 5030617 | ||
CX235072.1 | - | 4, 5 | 663 |