Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enoph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163035.1 | NP_001156507.1 | 6 | 947 | ||
NM_026421.3 | NP_080697.2 | 6 | 947 | |||
GenBank mRNA | AK009954.1 | - | 6 | 930 | ||
AK079062.1 | - | 6 | 941 | |||
AK080714.1 | - | 6 | 898 | |||
AK082747.1 | - | 6 | 922 | |||
AK152021.1 | - | 6 | 921 | |||
AK159356.1 | - | 6 | 920 | |||
AK167575.1 | - | 6 | 958 | |||
AK168696.1 | - | 6 | 816 | |||
BC021429.1 | - | 6 | 874 | 5030617 |