Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006419.2 | NP_006410.1 | 3 | 181 | ||
XM_006714063.3 | XP_006714126.1 | 2 | 174 | |||
GenBank mRNA | AF029894.1 | - | 2 | 137 | ||
AF044197.1 | - | 3 | 193 | |||
AJ002211.1 | - | 2 | 137 | |||
BC012589.1 | - | 2 | 153 | 4246053 | ||
BI836728.1 | - | 2 | 133 | |||
CR407624.1 | - | 2 | 103 |